5-Methylisoxazol-4-carboxylsyre ≥98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
L14628.06EA
1070
DKK
L14628.06
5-Methylisoxazol-4-carboxylsyre ≥98%
5-Methylisoxazol-4-carboxylsyre
Formel:
C₅H₅NO₃ Molekylvægt: 127,1 g/mol Smeltepunkt: 144…149 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00955655 CAS-nummer: 42831-50-5 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...