Experimental kit, magnetism and e-learning
Leverandør: EDUCATIONSCIENCE COLLECTION P
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
765-0284EA 0 DKK
765-0284
Experimental kit, magnetism and e-learning
Undervisningsmateriale Fysik undervisningsmateriale Electricity/Magnetism Learning Activities
Experimental set in Ellära and Magnetism, primarily intended for compulsory school upper secondary school or equivalent level of knowledge. The experiments can be performed as student experiments or demonstrations. For some experiments, 24 V AC is required. Appropriate power supply rectifier cube 765-1371. Special laboratory room is not a requirement. The kit includes everything needed to perform the experiments described in the tutorial, with the exception of some solutions such as sulfuric acid, sodium hydroxide, etc. The tutorial contains more than 50 experiments. Most qualitative. For some experiments, there is also a theory as a background to the experiments. The material is stored in a specially decorated wooden box. Through richness of detail and many combination possibilities, basic physics experiments can be performed in a number of different areas. Examples of experiments
  • Resistance and heat generation
  • Magnets, magnetic poles and power lines
  • Magnetic field around conductors and coils
  • Electromagnets (bell, telegraph)
  • Induction (generator, transformer)
  • DC and AC motors
  • Rotary coil and soft iron type instruments
  • Chemical effects of the current (elements, lead accumulator, electrolysis)
  • Thermoelectricity