Proteinase K (from Tritirachium album)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0706-100MGEA 568 DKK
VWRC0706-100MG VWRC0706-500MG
Proteinase K (from Tritirachium album)
A non specific protease with a molecular weight of approximately 18 kDa. Exhibits high stability and activity in the presence of SDS, EDTA, and urea, as well as over a wide pH range. Activity >30 units/mg.
  • No DNase or RNase detected
  • Lyophilised powder
Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range.