Electronic blood pressure monitor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-1981EA 1110 DKK
EDSCGM01022-B
Electronic blood pressure monitor
Measuring range: blood pressure 5,4 to 36 kPa (40 to 270 mmHg), pulse range 40 to 200 s/min
Precision: blood pressure: ±0,4 kPa (±3 mmHg), pulse rate within ≤ ±5% of reading value
Measuring method: automatically charging and measuring exhaust
Power supply: 4×1,5 AA batteries, steady 6 V/600 mA