Lux meter, digital 200,000Lux
Leverandør: EDUCATIONSCIENCE COLLECTION P
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761-0411EA 0 DKK
761-0411
Lux meter, digital 200,000Lux
Lysmåler
  • Format:
  • 2000 counts
  • Analogue bar chart
  • Automatic range and manual range
  • Automatic power off
  • Naximum and minimum
  • Relative measurements
  • Peak measurements
  • FC/Lux
  • Data hold
  • Display illumination
  • Low battery indicator