Comparative maxilla set for forensic and anthropological studies
Leverandør: Ward's
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761-0386EA 3740 DKK
WARD823573
Comparative maxilla set for forensic and anthropological studies
Discover forensic methods of identifying the race/ethnicity of an individual with this three-piece replica set. Differences in the maxillary, nasal, dental and palate structures among Asian, African, and European males are clearly illustrated.