Ballistics flashcards, Ward's®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761-0280EA 749 DKK
WARD323382
Ballistics flashcards, Ward's®
Observe lands and grooves around the entire circumference of bullets with flashcards. Images of four separate bullets are provided in order to make comparisons, in addition to images of the breach face of four shell casings, which show the firing pin impression on the primer of the casing.
  • Size: 7,6×12,7 mm
Emballering: Set of 20 flashcards.