Adhesive clips
Leverandør: Ward's
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WARD159790EA 146 DKK
WARD159790
Adhesive clips
Use clips to attach string to any vertical surface for ballistics determinations.
  • Removable and reusable
  • Pack of 10
The colourless adhesive wax backing on these clips will never dry out, so you can remove and reuse them repeatedly.