Vernier caliper
Leverandør: Ward's
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WARD154081EA 77.1 DKK
WARD154081
Vernier caliper
High impact plastic caliper.
Inside and outside measuring range of 140 mm