Ballistic angle set
Leverandør: Ward's
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761-0266EA 2130 DKK
761-0266 761-0264 761-0265
Ballistic angle set
Undervisningsmateriale Forensics Educational Materials Retsmedicinske sæt
Follow the path of a bullet from weapon to victim or surface using the components of this ballistics set. Two sets of pre-drilled plastic blocks simulate the holes produced by 9 mm and 0,45 calibre bullets entering at 30, 45, 60, and 90 degree angles. Mount the blocks on a wall or any surface and determine trajectory using either trajectory rods or string, available separately.
  • Simulate bullet holes for trajectory determinations