Magnetism school
Leverandør: EDUCATIONSCIENCE COLLECTION P
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
761-0161EA 1770 DKK
761-0161 761-0160
Magnetism school
Undervisningsmateriale Fysik undervisningsmateriale Electricity/Magnetism Learning Activities
Learn the principles of magnetism with these two activity sets designed for middle and high school students. Two sets of activities each with 10 experiments designed for students to work independently or in groups of 2. Each experiment is supplied with teacher and pupil laminated cards giving clear and instructions, in Swedish and with illustrations. The activity sets are designed so that the student can follow a theme through a series of experiments and discover the science whilst enjoying the experience.
Leveringsinformation: Examples of experiments included: Discovering magnetic fields using magnetic jewelry, assembling a simple electric motor, working with floating magnets and creating new graphic design shapes with magnetic powder.