Molecular models: DNA/RNA
Leverandør: Edvotek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
763-0004EA 586 DKK
763-0004 763-0005
Molecular models: DNA/RNA
Modeller Molekylærbiologi modeller
Students can build a model of DNA using this simple and colourful system.
  • Colour-coded parts to represent the purines, pyrimidines, deoxyribose and phosphate groups
  • Possibility to make up the double helix of DNA
  • Use together with the RNA protein synthesis kit to model translation and transcription
Suitable for modelling DNA replication.
Bestillingsoplysninger: Includes differently sized purines and pyrimidines. The correct number of hydrogen bonds and the minor and major grooves are shown.