Electrophoresis power supplies, EVT 300
Leverandør: Edvotek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
732-1424EA 2890 DKK
732-1424
Electrophoresis power supplies, EVT 300
Strømforsyninger
EVT 300 power supplies can operate M6Plus units, two M12 units or two HexaGels (at 75 V). The EVT 300 runs gels as fast as 20 to30 minutes (at 150 V).