(S)-(-)-Nicotin, Msynth® plus, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
L(-)-Nicotin
M-Clarity™ quality level = MQ200
8.45200.0100EA
0
DKK
8.45200.0100
8.45200.1000
(S)-(-)-Nicotin, Msynth® plus, Sigma-Aldrich®
(S)-(-)-Nicotin
Formel:
C₁₀H₁₄N₂ Molekylvægt: 162,23 g/mol Kogepunkt: 247 °C (1013 hPa) Smeltepunkt: –79 °C Massefylde: 1,01 g/cm³ (20 °C) Flammepunkt: >100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006369 CAS-nummer: 54-11-5 EINECS: 200-193-3 UN nummer: 1654 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...