VWR®, Mixed Anions Standard, 7 components in H₂O
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
91298.180EA 1430 DKK
91298.180
VWR®, Mixed Anions Standard, 7 components in H₂O
Standarder Anioniske standarder Multi-Element Anioinic Standards
7 components: 100 mg/l each of Chlorides (Cl-), Fluorides (F-), Sulphates (SO42-), Nitrates (NO3-), Nitrites (NO2-), Bromides (Br-), Phosphates (PO43-) in H₂O
Certificeringer: ISO 17025/17034