VWR®, Combined Five Anion Standard, 5 components in H₂O
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
91289.180EA 1210 DKK
91289.180
VWR®, Combined Five Anion Standard, 5 components in H₂O
Standarder Anioniske standarder Multi-Element Anioinic Standards
5 components: Fluorides (F-) 20 mg/l, Chlorides (Cl-) 30 mg/l, Nitrates (NO3-) 100 mg/l, Phosphates (PO43-) 150 mg/l, Sulphates (SO42-) 150 mg/l in H₂O
Certificeringer: ISO 17025/17034