VWR®, Sulphur in 5% Biodiesel (B5) with blank (2×100 ml)
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
91696.001EA 1050 DKK
91696.001
VWR®, Sulphur in 5% Biodiesel (B5) with blank (2×100 ml)
Standarder Kationiske standarder Sulphur Standards
100 ml of each: Sulfur 0,0015% w/w in Biodiesel 5; Sulfur in 5% Biodiesel (B5) Blank.
Advarsel: Irritant.