2-Chlorbenzensulfonylchlorid 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD9827-10GEA 179 DKK
BLDPBD9827-10G BLDPBD9827-25G BLDPBD9827-5G BLDPBD9827-1G
2-Chlorbenzensulfonylchlorid 96%
2-Chlorbenzensulfonylchlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄Cl₂O₂S
Molekylvægt: 211,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051520
CAS-nummer: 2905-23-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR