4-Hydroxy-3-nitropyridine-N-oxide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD97723-1GEA 497 DKK
BLDPBD97723-1G BLDPBD97723-250MG BLDPBD97723-5G
4-Hydroxy-3-nitropyridine-N-oxide 98%
4-Hydroxy-3-nitropyridine-N-oxide

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₄N₂O₄
Molekylvægt: 156,0963 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00234062
CAS-nummer: 31872-57-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR