4-Chloro-2-fluoro-5-nitrobenzoic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD95710-25GEA 450 DKK
BLDPBD95710-25G BLDPBD95710-5G BLDPBD95710-100G BLDPBD95710-10G
4-Chloro-2-fluoro-5-nitrobenzoic acid 97%
4-Chloro-2-fluoro-5-nitrobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₃ClFNO
Molekylvægt: 219,55 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115725
CAS-nummer: 35112-05-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR