4-Bromoisoxazole 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD95254-10GEA
1280
DKK
BLDPBD95254-10G
BLDPBD95254-5G
BLDPBD95254-250MG
BLDPBD95254-25G
BLDPBD95254-1G
4-Bromoisoxazole 97%
4-Bromoisoxazole
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formel:
C₃H₂BrNO Molekylvægt: 147,9600 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD02179569 CAS-nummer: 97925-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...