4-(Succinimidylcarboxy)benzophenone 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD94379-250MGEA
969
DKK
BLDPBD94379-250MG
BLDPBD94379-100MG
BLDPBD94379-1G
4-(Succinimidylcarboxy)benzophenone 98%
4-(Succinimidylcarboxy)benzophenone
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formel:
C₁₈H₁₃NO₄ Molekylvægt: 323,2995 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00058572 CAS-nummer: 91990-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...