4,5-Dimethylpyridazine-3,6-diol 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD92669-1GEA 117 DKK
BLDPBD92669-1G BLDPBD92669-25G BLDPBD92669-5G BLDPBD92669-10G
4,5-Dimethylpyridazine-3,6-diol 97%
4,5-Dimethylpyridazine-3,6-diol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₈N₂O₂
Molekylvægt: 140,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00067744
CAS-nummer: 5754-17-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR