1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD9266-100GEA 318 DKK
BLDPBD9266-100G BLDPBD9266-10G BLDPBD9266-5G BLDPBD9266-500G BLDPBD9266-25G
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₈H₂₄O₉
Molekylvægt: 504,5 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005357
CAS-nummer: 6974-32-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR