4,4,5,5,5-Pentafluorpentan-1-ol 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD91944-5GEA 225 DKK
BLDPBD91944-5G BLDPBD91944-100G BLDPBD91944-25G BLDPBD91944-1G BLDPBD91944-10G
4,4,5,5,5-Pentafluorpentan-1-ol 98%
4,4,5,5,5-Pentafluorpentan-1-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₇F₅O
Molekylvægt: 178,1 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00153224
CAS-nummer: 148043-73-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR