4'-(Trifluoromethoxy)acetanilide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD91686-5GEA 140 DKK
BLDPBD91686-5G
4'-(Trifluoromethoxy)acetanilide 97%
4'-(Trifluoromethoxy)acetanilide

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₈F₃NO₂
Molekylvægt: 219,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00075293
CAS-nummer: 1737-06-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR