4-Cyano-4'-hexyloxybiphenyl 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD91327-100GEA 5520 DKK
BLDPBD91327-100G BLDPBD91327-1G BLDPBD91327-5G BLDPBD91327-25G
4-Cyano-4'-hexyloxybiphenyl 98%
4-Cyano-4'-hexyloxybiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₂₁NO
Molekylvægt: 279,38 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134145
CAS-nummer: 41424-11-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR