2-Bromo-5-(4-morpholino)pyridine 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD90860-1GEA 109 DKK
BLDPBD90860-0.025G BLDPBD90860-1G BLDPBD90860-5G BLDPBD90860-25G
2-Bromo-5-(4-morpholino)pyridine 96%
2-Bromo-5-(4-morpholino)pyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₁BrN₂O
Molekylvægt: 243,1004 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02681930
CAS-nummer: 200064-11-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR