4-(4-Methoxybenzyl)piperidine hydrochloride 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD90659-25GEA 2450 DKK
BLDPBD90659-25G BLDPBD90659-5G BLDPBD90659-1G
4-(4-Methoxybenzyl)piperidine hydrochloride 95%
4-(4-Methoxybenzyl)piperidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₁₉NO·ClH
Molekylvægt: 241,757 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03840137
CAS-nummer: 37581-27-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR