4-(1H-Pyrazol-1-yl)benzenesulphonyl chloride 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD89559-1GEA 427 DKK
BLDPBD89559-1G BLDPBD89559-250MG BLDPBD89559-100MG BLDPBD89559-5G
4-(1H-Pyrazol-1-yl)benzenesulphonyl chloride 96%
4-(1H-Pyrazol-1-yl)benzenesulphonyl chloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₇ClN₂O₂S
Molekylvægt: 242,682 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02681937
CAS-nummer: 18336-39-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR