N-Cbz-D-phenylalaninol 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD8884-25GEA
876
DKK
BLDPBD8884-25G
BLDPBD8884-10G
BLDPBD8884-1G
BLDPBD8884-5G
N-Cbz-D-phenylalaninol 97%
N-Cbz-D-phenylalaninol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₇H₁₉NO₃ Molekylvægt: 285,3 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00191193 CAS-nummer: 58917-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...