5-Nitro-m-toluidine 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD88107-250MGEA 125 DKK
BLDPBD88107-250MG BLDPBD88107-1G BLDPBD88107-25G BLDPBD88107-100MG BLDPBD88107-5G
5-Nitro-m-toluidine 98%
5-Nitro-m-toluidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₈N₂O₂
Molekylvægt: 152,1500 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00082655
CAS-nummer: 618-61-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR