3-Iodobenzylamine hydrochloride 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD87519-250MGEA
62.1
DKK
BLDPBD87519-250MG
BLDPBD87519-1G
BLDPBD87519-25G
BLDPBD87519-5G
BLDPBD87519-10G
BLDPBD87519-0.025G
3-Iodobenzylamine hydrochloride 97%
3-Iodobenzylamine hydrochloride
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₉ClIN Molekylvægt: 269,5105 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012857 CAS-nummer: 3718-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...