Prostaglandin E1 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD8719-250MGEA 3360 DKK
BLDPBD8719-250MG BLDPBD8719-100MG BLDPBD8719-50MG BLDPBD8719-25MG
Prostaglandin E1 98%
Prostaglandin E1

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₀H₃₄O₅
Molekylvægt: 354,487 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00077860
CAS-nummer: 745-65-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR