3-Ethynylthiophene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD86719-10GEA 1100 DKK
BLDPBD86719-10G BLDPBD86719-5G BLDPBD86719-25G BLDPBD86719-1G
3-Ethynylthiophene 95%
3-Ethynylthiophene

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₄S
Molekylvægt: 108,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04039973
CAS-nummer: 67237-53-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR