3-Bromo-1H-indazole 97%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD85334-250MGEA
69.8
DKK
BLDPBD85334-250MG
BLDPBD85334-5G
BLDPBD85334-1G
BLDPBD85334-25G
BLDPBD85334-10G
3-Bromo-1H-indazole 97%
3-Bromo-1H-indazole
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₅BrN₂ Molekylvægt: 197,04 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00159926 CAS-nummer: 40598-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...