3-Amino-3-(2,4-dichlorophenyl)-propionic acid 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD83846-1GEA 714 DKK
BLDPBD83846-1G BLDPBD83846-250MG BLDPBD83846-100MG
3-Amino-3-(2,4-dichlorophenyl)-propionic acid 95%
3-Amino-3-(2,4-dichlorophenyl)-propionic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₉Cl₂NO₂
Molekylvægt: 234,079 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02663185
CAS-nummer: 152606-17-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR