3,5-Lutidine-N-oxide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD82886-1GEA 179 DKK
BLDPBD82886-1G BLDPBD82886-5G BLDPBD82886-25G BLDPBD82886-10G
3,5-Lutidine-N-oxide 98%
3,5-Lutidine-N-oxide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₉NO
Molekylvægt: 123,1525 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00126839
CAS-nummer: 3718-65-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR