3,5-Difluoro-4-formylphenylboronic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD82602-10GEA 2260 DKK
BLDPBD82602-0.025G BLDPBD82602-10G BLDPBD82602-25G BLDPBD82602-250MG BLDPBD82602-5G BLDPBD82602-1G
3,5-Difluoro-4-formylphenylboronic acid 95%
3,5-Difluoro-4-formylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅BF₂O₃
Molekylvægt: 185,9206 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07784344
CAS-nummer: 870718-11-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR