4-Acetyl-3-fluorophenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD8237-5GEA 1530 DKK
BLDPBD8237-5G BLDPBD8237-250MG BLDPBD8237-1G
4-Acetyl-3-fluorophenylboronic acid 98%
4-Acetyl-3-fluorophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₈BFO₃
Molekylvægt: 181,9600 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06796422
CAS-nummer: 481725-35-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR