2-Methoxy-5-pyrimidineboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD8107-100GEA 3900 DKK
BLDPBD8107-100G BLDPBD8107-250MG BLDPBD8107-10G BLDPBD8107-5G BLDPBD8107-25G BLDPBD8107-1G
2-Methoxy-5-pyrimidineboronic acid 98%
2-Methoxy-5-pyrimidineboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₇BN₂O₃
Molekylvægt: 153,9317 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03094664
CAS-nummer: 628692-15-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR