3-(tert-Butoxy)-6-chloropyridazine 97%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD80945-25GEA
4940
DKK
BLDPBD80945-25G
BLDPBD80945-1G
BLDPBD80945-5G
BLDPBD80945-250MG
3-(tert-Butoxy)-6-chloropyridazine 97%
3-(tert-Butoxy)-6-chloropyridazine
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₈H₁₁ClN₂O Molekylvægt: 186,6400 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD03425769 CAS-nummer: 17321-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...