2-Fluoro-5-(trifluoromethyl)phenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD8072-5GEA 272 DKK
BLDPBD8072-5G BLDPBD8072-100G BLDPBD8072-25G BLDPBD8072-1G
2-Fluoro-5-(trifluoromethyl)phenylboronic acid 98%
2-Fluoro-5-(trifluoromethyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BF₄O₂
Molekylvægt: 207,92 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03701538
CAS-nummer: 352535-96-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR