3,5-Dichloropyridine-2-carbonitrile 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD8052-5GEA 85.3 DKK
BLDPBD8052-5G BLDPBD8052-500G BLDPBD8052-10G BLDPBD8052-1G BLDPBD8052-100G BLDPBD8052-25G
3,5-Dichloropyridine-2-carbonitrile 97%
3,5-Dichloropyridine-2-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₂Cl₂N₂
Molekylvægt: 173,00 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788758
CAS-nummer: 85331-33-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR