3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD80247-1GEA 427 DKK
BLDPBD80247-1G BLDPBD80247-5G BLDPBD80247-250MG
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 98%
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₂₇BN₂O₃
Molekylvægt: 330,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05864308
CAS-nummer: 883738-38-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR