1-(2-Furoyl)piperazine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD7861-100GEA 2140 DKK
BLDPBD7861-100G BLDPBD7861-25G BLDPBD7861-5G BLDPBD7861-1G
1-(2-Furoyl)piperazine 98%
1-(2-Furoyl)piperazine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₂N₂O₂
Molekylvægt: 180,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00038831
CAS-nummer: 40172-95-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR