2-(Methylthio)benzo[d]oxazole 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD77392-25GEA 1050 DKK
BLDPBD77392-25G BLDPBD77392-5G BLDPBD77392-100G BLDPBD77392-1G
2-(Methylthio)benzo[d]oxazole 98%
2-(Methylthio)benzo[d]oxazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇NOS
Molekylvægt: 165,2100 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00160027
CAS-nummer: 13673-62-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR