4'-Methoxy-2-chloroacetophenone 98%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD7698-1GEA
256
DKK
BLDPBD7698-1G
BLDPBD7698-5G
4'-Methoxy-2-chloroacetophenone 98%
4'-Methoxy-2-chloroacetophenone
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₉H₉ClO₂ Molekylvægt: 184,6196 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00216508 CAS-nummer: 2196-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...