2-Methyl-3-(trifluoromethyl)benzyl bromide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD76820-250MGEA 101 DKK
BLDPBD76820-250MG BLDPBD76820-25G BLDPBD76820-1G BLDPBD76820-5G
2-Methyl-3-(trifluoromethyl)benzyl bromide 98%
2-Methyl-3-(trifluoromethyl)benzyl bromide

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈BrF₃
Molekylvægt: 253,06 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01631608
CAS-nummer: 261952-16-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR