3-Bromobenzaldehyde ethylene acetal 97% (by GC)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD7663-100GEA 1600 DKK
BLDPBD7663-100G BLDPBD7663-5G BLDPBD7663-1G BLDPBD7663-25G BLDPBD7663-0.025G BLDPBD7663-10G
3-Bromobenzaldehyde ethylene acetal 97% (by GC)
3-Bromobenzaldehyde ethylene acetal

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₉BrO₂
Molekylvægt: 229,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003209
CAS-nummer: 17789-14-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR